Whole Normal Kidney Tissue Lysate (Adult Normal)
Lieferant: Novus Biologicals
NB820-59231
NOVUNB820-59231EA
0
EUR
NOVUNB820-59231
Whole Normal Kidney Tissue Lysate (Adult Normal)
Zellen
Whole normal kidney tissue lysate (adult normal).
- Source: Human
Tested applications: Western blot, Immunoprecipitation. Tested reactivity: Human.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...