Human Recombinant Superoxide Dismutase 1 (from E. coli)
Lieferant: Novus Biologicals
NBC1-18544
NOVUNBC1-18544EA
654
EUR
NOVUNBC1-18544
Human Recombinant Superoxide Dismutase 1 (from E. coli)
Enzyme
The Recombinant Human SOD1/Cu-Zn SOD Protein is derived from E. coli. The Recombinant Human SOD1/Cu-Zn SOD Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
- Entrez Gene ID: 6647
Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...