Human recombinant enolase 1 (from E. coli)
Lieferant: Novus Biologicals
The Recombinant Human Enolase 1 Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Enolase 1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
- Entrez Gene ID: 2023
Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...