Anti-APLP2 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-32814
NOVUNBP1-32814EA 718 EUR
NOVUNBP1-32814
Anti-APLP2 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal APLP2 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.
Type: Primary
Antigen: APLP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human