Anti-ALIX rabbit polyclonal antibody

Lieferant: Novus Biologicals
NBP1-49701
NOVUNBP1-49701EA 742 EUR
NOVUNBP1-49701
Anti-ALIX rabbit polyclonal antibody
Antikörper
The Alix Antibody from Novus Biologicals is a rabbit polyclonal antibody to Alix. This antibody reacts with human. The Alix Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation.

Type: Primary
Antigen: Alix
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR