Anti-Aquaporin-7 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-54384
NOVUNBP1-54384EA 599 EUR
NOVUNBP1-54384
Anti-Aquaporin-7 Rabbit Polyclonal Antibody
Antikörper
The Aquaporin-7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aquaporin-7. This antibody reacts with human. The Aquaporin-7 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Aquaporin-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human