Anti-FBXO16 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-57614
NOVUNBP1-57614EA 595 EUR
NOVUNBP1-57614
Anti-FBXO16 Rabbit Polyclonal Antibody
Antikörper
The FBXO16 Antibody from Novus Biologicals is a rabbit polyclonal antibody to FBXO16. This antibody reacts with human. The FBXO16 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: FBXO16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR