Anti-LAMTOR2 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-71687
NOVUNBP1-71687EA 471 EUR
NOVUNBP1-71687
Anti-LAMTOR2 Rabbit Polyclonal Antibody
Antikörper
The ROBLD3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ROBLD3. This antibody reacts with human, mouse. The ROBLD3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:200

Type: Primary
Antigen: ROBLD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR