Anti-BRD4 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-86640
NOVUNBP1-86640EA 652 EUR
NOVUNBP1-86640
Anti-BRD4 Rabbit Polyclonal Antibody
Antikörper
The BRD4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BRD4. This antibody reacts with human. The BRD4 Antibody has been validated for the following applications: Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Simple Western

Type: Primary
Antigen: BRD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR