Anti-INPP5A Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-89361
NOVUNBP1-89361EA 652 EUR
NOVUNBP1-89361
Anti-INPP5A Rabbit Polyclonal Antibody
Antikörper
The INPP5A Antibody from Novus Biologicals is a rabbit polyclonal antibody to INPP5A. This antibody reacts with human. The INPP5A Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: INPP5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human