Anti-TBX2 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-89459
NOVUNBP1-89459EA 635 EUR
NOVUNBP1-89459
Anti-TBX2 Rabbit Polyclonal Antibody
Antikörper
The TBX2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TBX2. This antibody reacts with human, mouse. The TBX2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TBX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR