Anti-TNFSF11 Mouse Monoclonal Antibody (Alexa Fluor® 647)) [clone: 12A380]

Lieferant: Novus Biologicals
NBP2-27390
NOVUNBP2-27390EA 712 EUR
NOVUNBP2-27390
Anti-TNFSF11 Mouse Monoclonal Antibody (Alexa Fluor® 647)) [clone: 12A380]
Antikörper
Type: Primary
Antigen: TNFSF11 (tumor necrosis factor (ligand) superfamily, member 11)
Clonality: monoclonal
Clone: 12A380
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR