Anti-BAG3 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP2-27398
NOVUNBP2-27398EA 583 EUR
NOVUNBP2-27398
Anti-BAG3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BAG3 (BCL2-associated athanogene 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse