Anti-DDX58 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP2-27419
NOVUNBP2-27419EA 434 EUR
NOVUNBP2-27419
Anti-DDX58 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDX58 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 58)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR