EPA vial convenience kits, assembled
Lieferant: THERMO NATIONAL SCIENTIFIC
B7921
NSCAB7921EA 209 EUR
NSCAB7921
EPA vial convenience kits, assembled
Vials
Thermo Scientific™ national EPA vial convenience kits are designed for discrete water sampling applications.
Kits come with caps, septa and a certificate of analysis.