Precrimped, crimp-top vials, large opening, 11 mm
Lieferant: THERMO NATIONAL SCIENTIFIC
C4011-2WCV
NSCAC4011-2WCVEA 86.3 EUR
NSCAC4011-2WCV
Precrimped, crimp-top vials, large opening, 11 mm
Vials
Glass vials are precrimped with an aluminium seal and PTFE/red rubber septum.
  • Aluminium seal is crimped onto the vial following a purge with dry nitrogen
  • Sample is added via injection through the septum
Save time during sample preparation and reduce the risk of contamination.
Verpackung: Packaged in convenient vial trays with clear covers.