Vial crusher, national
Lieferant: THERMO NATIONAL SCIENTIFIC
C4040-VC
NSCAC4040-VCEA 12720 EUR
NSCAC4040-VC
Vial crusher, national
Ampullenöffner
Breaks and separates liquid from all 1 ml to 100 ml vials and ampoules.
  • Capable of processing approximately 5000 of 2 ml vials per hour
  • Reduces overall waste disposal costs by reducing the total volume of waste
Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapours. Environmentally responsible waste handling, crushed glass is automatically separated from solvent for separate disposal.