γ-Linolensäuremethylester
Lieferant: NU-CHEK-PREP
Synonyme:
Methyl-(6Z,9Z,12Z)-6,9,12-octadecatrienoat, Methyl-γ-linolenat, cis,cis,cis-6,9,12-Octadecatriensäuremethylester, 6Z,9Z,12Z-Octadecatriensäuremethylester, Methyl-cis,cis,cis-6,9,12-octadecatrienoat
GLC-411.0001
NUCKGLC-411.0001EA
0
EUR
NUCKGLC-411.0001
γ-Linolensäuremethylester
γ-Linolensäuremethylester
Formel:
C₁₉H₃₂O₂ Molecular Weight: 292,46 g/mol |
MDL:
MFCD00010455 CAS-Nummer: 16326-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...