Tristearin (Glycerintristearat)

Lieferant: NU-CHEK-PREP

Synonyme: Propane-1,2,3-triyl tristearate, Tristearin

T-160
NUCKT-160EA 0 EUR
NUCKT-160
Tristearin (Glycerintristearat)
Tristearin (Glycerintristearat)
Formel: C₅₇H₁₁₀O₆
Molecular Weight: 891,4797 g/mol
MDL: MFCD00036230
CAS-Nummer: 555-43-1
EINECS: 209-097-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR