Barcode reader, Nunc™
Lieferant: Thermo Fisher Scientific
330050
NUNC330050EA 598 EUR
NUNC330050
Barcode reader, Nunc™
Barcode- und RFID-Systeme
This durable barcode reader is ideal for simple track sampling. It is for use with any USB slot.
  • 'Plug and Play' function can automatically switch to read codes: 128A, 128B, 128C, 39 and interleave ²/₅ Convenient and easy to use with an Excel spread sheet
  • Scanner is a contact scanner which means it must be in contact with the barcode to read