Conical tubes, bulk, sterile, Thermo Scientific™
Lieferant: Thermo Fisher Scientific
332265 332270
NUNC332265EA 174 EUR
NUNC332265 NUNC332270
Conical tubes, bulk, sterile, Thermo Scientific™
Röhrchen Zentrifugenröhrchen
The Thermo Scientific™ Conical Centrifuge Tubes are sterile and ideal for applications requiring larger sample volumes.
  • Manufactured with high quality virgin resins
  • Sterilised by gamma irradiation per ISO 11137-2
  • Non-pyrogenic, non-cytotoxic
  • DNase/Rnase, BSE/TSE and human DNA free
Standards: Compliant with California Prop 65