Ölrot O 0,5% in Propylenglykol, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

O1516-250ML O1516-500ML
O1516-250MLEA 367 EUR
O1516-250ML O1516-500ML
Ölrot O 0,5% in Propylenglykol, Sigma-Aldrich®
Ölrot O

Oil red O is a lysochrome (fat-soluble dye) diazo dye used for staining of neutral triglycerides and lipids on frozen sections and some lipoproteins on paraffin sections. In histology, a supersaturated solution of oil red O in isopropanol may be used to stain fat in tissue.

Formel: C₂₆H₂₄N₄O
Lagertemperatur: Room Temperature
MDL: MFCD00003898
CAS-Nummer: 1320-06-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db861df49353969f31bdacf110373f0af69e40e&vid=8127ea78128711195b44982188087011024fd2c3&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance
Dark Red, Sediment Free
Conforms
UV/VIS Absorbance
Lambda Max 506 to 520 nm
0.40 - 1.00
Lambda Max 506 - 520 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR