Supelco® Organic Reference Standard, F.A.M.E. Mix RM-4
Lieferant: Merck
O7381-1AMP
O7381-1AMPEA 60.9 EUR
O7381-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix RM-4
Standards Organische Standards
Palmitic acid methyl ester (C16:0) 11,0% Stearic acid methyl ester (C18:0) 3,0% Oleic acid methyl ester (C18:1cis-9) 80,0% Linoleic acid methyl ester (C18:2cis-912) 6,0%.