Organo/Limonene Mount™

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

O8015-30ML O8015-100ML
O8015-30MLEA 102 EUR
O8015-30ML O8015-100ML
Organo/Limonene Mount™
Organo/Limonene Mount™

Organo/Limonene Mount™ is a ready-to-use mounting medium is made with limonene, a natural product from orange peels. It is good for coverslipping stained tissues and cell smears that can be dehydrated with organic solvents.


This mounting medium is suitable for organic solvent resistant immunohistochemistry (IHC) chromogens as well as an excellent choice for mounting H and E stained slides.


Bitte beachten Organo/Limonene Mount is a trademark of ImmunoBioScience Corp.

Flammpunkt: 48 °C
UN: 2052
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR