Block heater, digital, with heated lid

Lieferant: OHAUS
30392103
OHAU30392103EA 1535 EUR
OHAU30392103
Block heater, digital, with heated lid
Blockthermostate Blockthermostate, digital
These digital dry block heaters with heated lid are designed for applications that require repeatable results and superior temperature stability. These multi-purpose units are ideal for isothermal incubation, enzyme reactions, immunoassays, nucleic acid denaturation and a wide variety of other laboratory procedures.

  • Uniformity ±0,1 °C at 37 °C within the block
  • Hot warning symbol light is illuminated when the temperature is above 40 °C
  • Overshoot protection ensures unit will automatically stop heating if the unit exceeds the set temperature by 10 °C
  • Audible alarm sounds when time reaches zero and when unit reaches set-point temperature
  • Temperature calibration mode allows user to calibrate unit to an external temperature device

Integral support rod holder with locking knob accepts optional external temperature probe kit. Optional external temperature probe kit monitors actual block or sample temperature. Block heater accepts one microplate block or two separate interchangeable modular blocks, accommodating various tube sizes from 0,2 ml microtubes to test tubes or vials up to 85 mm in height. The heated lid helps to regulate the temperature and reduce the amount of condensation on sample lids.

Each block has a thermometer well for measuring block temperature. Efficient heating due to close tube and block contact with a low density design for exceptional temperature uniformity. Heaters require modular heating blocks for operation.

PID temperature control, with optional external RTD probe, offers a temperature stability as low as ±0,1 °C with a temperature uniformity as low as ±0,1 °C. Samples are heated to temperature quickly and accurately. Temperature is adjusted in ±0,1 °C increments.

Touch pad controls with easy-to-read, independent LED displays for temperature, and time. It provides repeatable and accurate results every time and is easily visible across lab benches. Shows set-point and actual temperature. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.

Units can be run in conditions from 18 to 33 °C, 20 to 80% relative humidity, non-condensing.

Bestellinformation: Modular heating blocks must be ordered separately.

Lieferumfang: Units include a detachable, 3-wire cord and plug.

Bitte beachten To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR