Defender™ 6000 Hybrid Bench Scales
Lieferant: OHAUS
30575550 30575551 30575552 30575553 30575554 30575555 30612473 30612474 30612475 30612476 30612477 30612498
OHAU30575550EA 2205 EUR
OHAU30575550 OHAU30575551 OHAU30575552 OHAU30575553 OHAU30575554 OHAU30575555 OHAU30612473 OHAU30612474 OHAU30612475 OHAU30612476 OHAU30612477 OHAU30612498
Defender™ 6000 Hybrid Bench Scales
Waagen
Defender 6000 stands up to harsh cleaning in processing and packaging environments with IP68 immersion/ IP69K washdown protection, 316 stainless steel construction and hermetically-sealed load cell.
  • 6-digit, 7-segment LCD display with white backlight, 45 mm high digits
  • 6 D (LR20) Batteries (Included)
  • i-DT61PW indicator
Versatile scales include fast weight display and multiple software modes to meet food, chemical processing, and packaging needs.
150% overload capacity protection, 3-colour check light bar, menu and key lock, selectable environmental and auto-print settings, stability indicator, overload/underload indicators, low battery indicator, auto shut-off, auto tare, adjustable rubber feet.
316 stainless steel platform and frame, hermetically-sealed stainless steel load cell, IP68/IP69K food grade polycarbonate housing indicator and 316 stainless steel mounting bracket.
Weighing, counting with APW optimizsation, percent weighing, checkweighing, checkcounting,
dynamic weighing/display hold.
Standards: NSF certified.