LED fluorescence excitation light source for CKX, BX, IX, SZX and MVX microscopes with broad spectrum, high power white illumination. Excitation wavelengths from 365-635nm (DAPI Cy5). Compatible with the narrow 365 nm DAPI filter sets. Global light 1 * 1 ST
Lieferant: OLYMPUS OPTICAL
LED fluorescence excitation light source for CKX, BX, IX, SZX and MVX microscopes with broad spectrum, high power white illumination. Excitation wavelengths from 365-635nm (DAPI Cy5). Compatible with the narrow 365 nm DAPI filter sets. Global light 1 * 1 ST