E-Z Select® Robotic Reservoir, 600 ml
Lieferant: OMEGA BIO-TEK
PS1-600V-14
OMEGPS1-600V-14EA 216 EUR
OMEGPS1-600V-14
E-Z Select® Robotic Reservoir, 600 ml
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The E-Z Select® Robotic Reservoir, 600 ml is designed for holding large volume reagent for use on automated platforms such as Hamilton Microlab STAR.
  • Large capacity: 600 ml reagent reservoir
  • Automatable: ANSI/SLAS compliant footprint for compatible automated platforms
  • Optimised design: V-bottom indentations ensure adequate aspirating and mixing
The E-Z Select® Robotic Reservoir, 600 ml is designed for holding large volume reagent for use on automated platforms such as Hamilton Microlab STAR. The reservoir is ANSI/SLAS compliant and able to fit on any accommodating deck space. The bottom of the reservoir incorporates V-bottom indentations to allow single channel and 96 probe heads to perform adequate aspirating and mixing. The maximum working volume is 550 ml and maximum fill volume is 600 ml. The height of the reservoir is 90,6 mm.