Two-way adaptors, PEEK™
Lieferant: OMNIFIT
002306
OMNI002306EA 46.9 EUR
OMNI002306
Two-way adaptors, PEEK™
Fittings und Verbinder
Omnifit® threaded adaptors are a useful range of problem solving parts that enable to convert thread sizes quickly and easily, accepting a ¹/₄"-28 UNF male fitting at one end and a range of male and female thread types at the other.