CMOS cooled colour camera for fluorescence applications with high sensitivity and low noise.
- Peltier-cooled to 45 °C below camera body temperature
- C-mount connection for microscopes
- High-speed live view camera for smooth results at maximum resolution
- Extremely reliable colour fidelity sensor
- High resolution and long time exposure for vivid details
Recommended for routinary and research level on different observation methods.
Bestellinformation: Includes a comprehensive software suite.
Lieferumfang: Equipped with a calibration slide for establishing measurement baselines, USB cable and power supply for cooling system.