Watertight Hard Case, CS-01
Lieferant: OPTIKA
CS-01
OPTKCS-01EA 224 EUR
OPTKCS-01
Watertight Hard Case, CS-01
Beutel Equipment Bags and Instrument Cases
CS-01 is a watertight hard case, which is made to offer superior protection from the elements, water, dust and impacts when transporting fragile and valuable objects, like a microscope.
Convoluted foam in the lid and cubed foam (three layers) in the bottom with pick and pluck system to easely adapt the case to different microscopes.
It can be used for carrying following models:
– B-190 Series: B-191PL, B-192PL, B-193PL
– B-290 Series: B-292, B-292PLI, B-293, B-293PLI
– B-380 Series: B-382. B-382PLI, B-383, B-383PLI, B-382PH, B-382PHI, B-383PH, B-383PHI, B-383DK
– B-510 Series: B-510BF, B-510Ergo, B-510PH, B-510DK
– SLX Series: SLX-1, SLX-2, SLX-3
– SZ Series: all models equipped with bases SZ-ST1, SZ-ST2, SZ-ST3, SZ-ST7, SZ-ST8
Standards: IP 67, ATA 300 and military certified.