Anti-EPN2 Mouse Monoclonal Antibody [clone: 1F1]

Lieferant: OriGene
CF504298
ORIGCF504298EA 833 EUR
ORIGCF504298
Anti-EPN2 Mouse Monoclonal Antibody [clone: 1F1]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: EPN2
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR