Human Tagged ORF Clone, PSMA (FOLH1) (NM_001014986)
Lieferant: OriGene
FOLH1 (Myc-DDK-tagged)-Human folate hydrolase (prostate-specific membrane antigen) 1 (FOLH1), transcript variant 2.
This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene. The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10 µg of transfection-ready, dried plasmid DNA (reconstitute with 100 µl of water).
Reconstitution:
Centrifuge at 5000×g for 5 min
Carefully open the tube and add 100ul of sterile water to dissolve the DNA
Close the tube and incubate for 10 minutes at room temperature
Briefly vortex the tube and then do a quick spin (less than 5000×g) to concentrate the liquid at the bottom
Store the suspended plasmid at −20 °C, the DNA is stable for at least one year from date of shipping when stored at −20 °C
Synonyms: FGCP; FOLH; GCP2; GCPII; mGCP; NAALAD1; NAALAdase; PSM; PSMA
E. coli selection: Kanamycin (25 µg/ml)
Mammalian cell selection: Neomycin
Sequence data: ORF nucleotide sequence and protein sequence
Restriction sites: SgfI-MluI
ACCN: NM_001014986
ORF size: 2157 bp
UniProt ID: Q04609
Cytogenetics: 11p11.12
Protein families: Druggable genome, protease, transmembrane
Molecular weight: 80,6 kDa
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...