Human Untagged LRRC15 Clone
Lieferant: OriGene
SC305914
ORIGSC305914EA 1130 EUR
ORIGSC305914
Human Untagged LRRC15 Clone
Vektoren, Plasmide und Bibliotheken
LRRC15 (untagged)-Human leucine rich repeat containing 15 (LRRC15), transcript variant 2.
E. coli selection: Kanamycin (25 µg/ml)
Mammalian cell selection: Neomycin
Insert size: 1700 bp