Human Untagged FBXL17 Clone
Lieferant: OriGene
SC327180
ORIGSC327180EA
982
EUR
ORIGSC327180
Human Untagged FBXL17 Clone
Vektoren, Plasmide und Bibliotheken
FBXL17 (untagged)-Human F-box and leucine-rich repeat protein 17 (FBXL17). The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10 µg of transfection-ready, dried plasmid DNA (reconstitute with 100 µl of water).
- Centrifuge at 5000×g for 5 min
- Carefully open the tube and add 100 µl of sterile water to dissolve the DNA
- Close the tube and incubate for 10 minutes at room temperature
- Briefly vortex the tube and then do a quick spin (less than 5000×g) to concentrate the liquid at the bottom
- Store the suspended plasmid at −20 °C. The DNA is stable for at least one year from date of shipping when stored at −20 °C
E. coli Selection: Kanamycin (25 µg/ml)
Synonyms: Fbl17; Fbx13; FBXO13
Mammalian cell selection: Neomycin
Tag: Tag free