Human Recombinant AFT2 (from sf9 cells)
Lieferant: OriGene
TP710017
ORIGTP710017EA
715
EUR
ORIGTP710017
Human Recombinant AFT2 (from sf9 cells)
Proteine und Peptide
Recombinant protein of human activating transcription factor 2 (ATF2), full length, with C-terminal polyhistidine tag, expressed in sf9 cells.
Buffer: 50 mM Tris-HCl, pH 8,0, 150 mM NaCl, 20% glycerol