Electrolyte solution for polarographic dissolved oxygen probes, Orion™
Lieferant: Thermo Orion
080514
ORIO080514EA 28.4 EUR
ORIO080514
Electrolyte solution for polarographic dissolved oxygen probes, Orion™
Elektrolyte Elektrodenfülllösungen
This electrolyte solution helps to maintain DO probes in optimal working condition.