Resistor kit for conductivity meters, Orion™
Lieferant: Thermo Orion
1010001
ORIO1010001EA 1020 EUR
ORIO1010001
Resistor kit for conductivity meters, Orion™
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
This resistor calibration kit can be used to verify the performance of Thermo Scientific™ Orion™ Versa Star conductivity meters, Thermo Scientific™ Orion™ Star A series conductivity meters and legacy Orion™ Star series conductivity meters.
  • Conductivity calibration resistor kit with 8 pin MiniDIN connection
Lieferumfang: Kit includes six resistors that simulate nominal conductance values between 1 µS and 100 mS.