Orion™ Conductivity Standards and Solutions, Thermo Scientific
Lieferant: Thermo Orion
990106 011001 011006 01100910 11007 11008 O1100510 O1100710
663-0044EA 152 EUR
663-0044 ORIO011001 663-0022 ORIO01100910 ORIO11007 ORIO11008 ORIOO1100510 ORIOO1100710
Orion™ Conductivity Standards and Solutions, Thermo Scientific
Standards Leitfähigkeits-Standards
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
  • For use with Thermo Scientific™ Orion™ Conductivity Probes
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.