pH/mV/°C/ISE meter, bench, Orion™ Dual Star™

Lieferant: Thermo Orion
2115000 2115001 2115101 2115102 2115205 2115203 2115202 2115204 2115201
ORIO2115000EA 2090 EUR
ORIO2115000 ORIO2115001 ORIO2115101 ORIO2115102 ORIO2115205 ORIO2115203 ORIO2115202 ORIO2115204 ORIO2115201
pH/mV/°C/ISE meter, bench, Orion™ Dual Star™
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
These bench top meters with two measuring channels are designed for accurate and reliable pH, ion concentration, mV, ORP and temperature measurements. Ideal for high throughput pH and ion analysis in the lab.

  • Numeric keypad and menu-specific function keys; split screen display for viewing two measurements at the same time
  • Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
  • Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
  • Non volatile meter memory holds up to 1000 data points with time and date stamp
  • Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
  • Stir samples with ease using the meter-controlled Orion Star stirrer probe (included in some kits)
  • Meter-attachable electrode stand assists with moving and maintaining electrodes

Use the two channels to measure a pH sample and ISE sample, two pH samples or two ISE samples simultaneously or view each channel separately.

IP54-rated housing handles splashes and is wall-mountable to save space.

Double your measurement capabilities with the Thermo Scientific™ Orion™ Dual Star™ pH and ISE Benchtop Meter. Record accurate and reliable pH, ion concentration, mV, ORP and temperature measurements on two available meter channels, ideal for high throughput pH and ion analysis in the lab. Perform up to 6 point pH and ISE calibrations, log up to 1000 data point sets with time/date stamp and easily transfer to a printer or computer. Use the two channels to measure a pH sample and ISE sample, two pH samples or two ISE samples simultaneously or view each channel separately.

Isopotential point : 7,000 pH (adjustable), 1,000 ISE (adjustable)
AC adapter : Universal 50 to 60 Hz, 100 to 240 VAC power adapter (Cat. No. 1010003)
Relative humidity range : 5 to 85 %, non-condensing
Electrical requirements : 90 to 260 VAC 50/60 Hz

3 year meter warranty with adjustable backlight option. It has smart stability and smart averaging.

Standards: CE, TUV 3-1, FCC Class A.

Lieferumfang: Kits are supplied with electrode(s), electrode stand, universal power adapter, literature, computer interface cable and meter test certificate.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR