Test kit, pH for pure waters, Orion™
Lieferant: Thermo Orion
Kit provides fast, accurate pH results in high purity and low ionic strength waters.
- Increases measurement accuracy
- Reduces electrode drift
- Eliminates contamination of sample
Minimises problems encountered when measuring the pH of pure waters. Kit utilises the pure water pH additive pHISA® Adjustor to increase ionic strength. Net effect of the pH additive is made negligible by incorporating a special set of diluted buffers containing the same background of pHISA® Adjustor as the sample. A glass electrode is recommended for the measurement.
Recommended for samples such as well water, rain water, distilled/deionised water, boiler feed water and process water.
Lieferumfang: Ordering Information: Kit includes 4×475 ml bottles of pH buffer A (pH 6,97), 2×475 ml bottles of pH buffer B (pH 4,10), 2×50 L bottles of pHISA® Adjustor, one 1ml syringe, one holding tray, instruction card, and application procedure pamphlet.