Test kits and tablets for AQUAfast II colorimeters
Lieferant: Thermo Orion
Orion™ AQUAfast tablet reagents perform colorimetric measurements of common water parameters with convenient, rapidly dissolving tablets.
- For use with ORION™ AQUAfast AQ4000, ORION™ AQUAfast AQ3070, ORION™ AQUAfast AQ3700, and discontinued ORION™ AQ2 series meters and other colorimeters
- 50 to 100 tests per kit
Use Orion™ AQUAfast Tablet Reagents with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging. They are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide, and zinc.