Test kits in ampoules, AQUAfast™ IV reagents, Orion™
Lieferant: Thermo Orion
AC4004
AC4005
AC4006
AC4007
AC4011
AC4012
AC4016
AC4017
AC4046
AC4048
AC4055
AC4060
AC4065
AC4070
AC4078
AC4082
AC4095
AC4099
ORIOAC4004EA
211
EUR
ORIOAC4004
ORIOAC4005
ORIOAC4006
705-0751
ORIOAC4011
ORIOAC4012
ORIOAC4016
ORIOAC4017
ORIOAC4046
ORIOAC4048
ORIOAC4055
ORIOAC4060
ORIOAC4065
705-0752
ORIOAC4078
ORIOAC4082
705-0753
ORIOAC4099
Test kits in ampoules, AQUAfast™ IV reagents, Orion™
Umwelt-Prüfkits
Orion™ AQUAfast™ Auto-Test Ampoule Reagents can be used with the Orion™ AQUAfast™ AQ4000 colorimeter to simplify sample preparation and tests.
- Consistency and reproducibility improved with AQUAfast™ ampoules and an AQ4000 meter
- Auto-ID™ eliminates measurement errors
- Designed for use with colorimetric water and wastewater testing equipment
The Auto-Test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.