Total Nitrogen test kit for AQUAfast 3700 Colorimeter and AquaMate spectrophotometers
Lieferant: Thermo Orion
Orion™ AQUAfast™ Total Nitrogen reagents in digestion tubes are designed for use with Orion™ AQUAfast™ 3700 colorimeter and Orion™ AquaMate Vis and UV-Vis spectrophotometers. Just add the sample to the digestion tube, digest sample in thermoreactor and measure.
- Reagents are fully prepared and offer convenience and added safety for user
Digestion tubes contain the precise amount of reagent, are made of special optical glass, and fit the Orion™ AQUAfast™ COD165 thermoreactor.