Handheld turbidity meter, Orion™ AQUAfast™ AQ4500
Lieferant: Thermo Orion
AQ4500
ORIOAQ4500EA 1810 EUR
ORIOAQ4500
Handheld turbidity meter, Orion™ AQUAfast™ AQ4500
Trübungsmessgeräte
The ORION AQUAfast AQ4500 can provide the nephelometric and ratiometric principles of turbity measurement.
  • Log data into internal memory, or use the optional RS232 cable to send data to printer or download to a computer for easy time and date stamped results storage
  • Nephelometric and ratiometric measurements with autoranging
  • Dual LED light sources and drinking water/wastewater methods
Standards: ISO 7027, EPA 180.1, ASBC, EBC.
Bestellinformation: Includes seven glass vials, user guide and carrying case. Silicone oil and cloth set is available separately.