Benchtop meter-attached electrode stand, Orion™ Star A series
Lieferant: Thermo Orion
STARA-BEA
ORIOSTARA-BEAEA 163 EUR
ORIOSTARA-BEA
Benchtop meter-attached electrode stand, Orion™ Star A series
Elektrodenhalter
This meter-attached electrode stand can be used to easily place sensors in samples.
  • Simplifies rinsing and storage between samples
  • Cable management system minimises electrode breakage
  • 360° motion allows electrodes to be moved vertically, horizontally and front-to-back positioning into beakers or sample containers
Can be attached to either side of Thermo Scientific™ Orion™ Versa Star benchtop meters and Thermo Scientific™ Orion™ Star A210 and Star A110 series benchtop meters. Can also be used with weighted base for compatibility with any meter system (portable or benchtop).
Bestellinformation: The weighted base needs to be ordered separately.
Lieferumfang: Includes meter bracket, arm and electrode holder.