The Thermo Scientific™ Orion Star™ T910 pH titrators are designed to increase your laboratory productivity by automating titrations. Our core electrochemistry technology is integrated with a state-of-the-art reagent dispensing system to create modern, simplified automated titrators designed to make performing titrations easier, more reliable and more reproducible than manual titrations.
- Accurate, reproducible results–automating the titration workflow enables analysis to be run using the same setup parameters and calculations every time
- Fast setup time–setup and save electrode, titrant and titration parameters once using methods
- Prioritize lab throughput–start the titration, and let the instrument run unattended
- Minimal cost and downtime–easily replace titrator parts to minimise maintenance downtime while long lasting electrodes may reduce the cost of ownership
- Securely document test results–view results live, in the summary table and within the 100-point data log
- Applications database–tested and validated preprogrammed methods can be easily loaded onto your titrator via USB flash drive
- Equivalence point (1 or 2 points) or preset endpoint (1, 2 or 3 points) titration technique
- Up to 5 cycles per titration run with option to exclude cycles from average results and RSD calculations
Pair the Orion Star™ T910 pH titrator with any premium ROSS pH electrodes, select a preconfigured titrator and electrode kit.
The 8102BNUWP ROSS Ultra pH electrode and 927007MD stainless steel ATC temperature probe are ideal for most standard pH titrations and preset endpoint titrations that include temperature.
The 8172BNWP ROSS Sure-Flow electrode is designed for challenging samples that can clog standard pH electrodes or that require a fast-flowing sleeve-type junction.
Standards: CE, TUV 3-in-1, FCC, EN/EIC61326-1, IEC 61010, IP-51.
Zubehörinformationen: Expand the functionality of your Orion Star™ T910 pH titrator with key accessories designed to help increase your lab productivity. Easy to install replacement parts minimise maintenance downtime and ensure your Orion Star™ T900 Series titrators are ready for sample analysis.
Bestellinformation: Orion Star™ T910 pH titrator includes 20 ml burette, stirrer probe, dispenser probe, standard tubing kit, 1 L plastic bottle, GL 38 bottle cap with drying tube, computer cable, literature on USB drive, 110 to 240 V power supplies.
Orion Star™ T910 pH titrator standard ROSS™ kit includes 8102BNUWP ROSS Ultra™ pH electrode, 927007MD ATC probe, 20 ml burette, stirrer probe, dispenser probe, standard tubing kit, 1 L plastic bottle, GL38 bottle cap with drying tube, computer cable, literature on USB drive, 110 to 240 V power supplies.
Orion Star™ T910 pH titrator Sure-Flow™ ROSS™ kit includes 8172BNWP ROSS Sure-Flow ™ pH electrode, 927007MD ATC probe, 20 ml burette, stirrer probe, dispenser probe, standard tubing kit, 1 L plastic bottle, GL 38 bottle cap with drying tube, computer cable, literature on USB drive, 110 to 240 V power supplies.