Orion Star™ T920 Redox Titrators
Lieferant: Thermo Orion
START9200 START9201
ORIOSTART9200EA 3720 EUR
ORIOSTART9200 ORIOSTART9201
Orion Star™ T920 Redox Titrators
Titratoren
Use the Orion Star T920 redox titrator for dedicated redox titrations including sulphite/SO₂ and reducing sugar in juice and wine, ascorbic acid (vitamin C) and peroxide value in food products, dissolved oxygen in wastewater by Winkler titration and organic matter in soil.
  • Offer easy-to-use navigation from setup to live titration analysis to data transfer
  • Showcase titrations, onscreen instructions and help menus on a large color graphic touchscreen display
  • Create and save up to ten user-defined methods
  • Save up to 100 titration data sets with date/time stamp
  • Facilitate data transfer to printer, computer or USB drive
  • Equivalence point, preset endpoint (1, 2 or 3 points) or multiple known addition (titration techniques)
Titration techniques include equivalence point titrations and preset mV endpoint titrations for versatile sample analysis. Pair the titrator option with any premium Orion redox electrodes or select a preconfigured titrator and electrode kit.
Accurate, reproducible results-automating the titration workflow enables analysis to be run using the same setup parameters and calculations every time. Fast setup time-setup and save electrode, titrant and titration parameters once using methods. Prioritise lab throughput-start the titration, and let the instrument run unattended.
Minimal cost and downtime-easily replace titrator parts to minimize maintenance downtime while long lasting electrodes may reduce the cost of ownership. Securely Document Test Results–View results live, in the summary table and within the 100-point data log. Applications database-tested and validated preprogrammed methods can be easily loaded onto your titrator via USB flash drive.
Standards: CE, TUV 3-in-1, FCC, EN/EIC61326-1, IEC 61010, IP-51.
Lieferumfang: Orion Star T920 Redox titrator, includes 20 ml burette, stirrer probe, dispenser probe, standard tubing kit, 1 L plastic bottle, GL38 bottle cap with drying tube, computer cable, literature on USB drive, 110 to 240 V power supplies. Orion Star T920 redox titrator kit, includes 9778BNWP glass-body redox electrode, 20 ml burette, stirrer probe, dispenser probe, standard tubing kit, 1 L plastic bottle, GL 38 bottle cap with drying tube, computer cable, literature on USB drive, 110 to 240 V power supplies.