pH / ISE measurement module for Orion™ Versa Star Pro™ meters

Lieferant: Thermo Orion
VSTAR-ISE
ORIOVSTAR-ISEEA 993 EUR
ORIOVSTAR-ISE
pH / ISE measurement module for Orion™ Versa Star Pro™ meters
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This measurement module can be used to customise the measurement capabilities of the Orion™ Versa Star Pro™ meters. It adds pH, ion concentration, mV, ORP and temperature measurement capabilities to the meter.

  • Each Orion™ Versa Star Pro meter has four channels that can be customised with interchangeable measurement modules
  • Measurement modules are interchangeable
  • Up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Perform up to a six point ISE calibration with selectable units of ppm, M, mg/L, %, ppb or none
  • Incremental techniques include single known addition, double known addition, single known subtraction, double known subtraction, analate addition and analate subtraction
  • Advanced ISE features include non linear selectable auto-blank, low range stability and electrode ID with calibration verification
  • Customise measurements to show calibration history and pH electrode condition or simplify to show an extra large pH or ion concentration reading

Range (pH): −2,000 to +20,000
Range (mV): ±2000,0
Range (Temperature): −5,0 to +105,0 °C
Range (ISE): 0,0001 to 19990 with selectable units of ppm, Molar, mg/L, %, ppb or none

3-year replacement warranty.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR