pH measurement module for Orion™ Versa Star Pro™ meters
Lieferant: Thermo Orion
VSTAR-PH
ORIOVSTAR-PHEA 542 EUR
ORIOVSTAR-PH
pH measurement module for Orion™ Versa Star Pro™ meters
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This measurement module can be used to customise the measurement capabilities of the Orion™ Versa Star Pro™ meters. It adds pH, mV, ORP and temperature measurement capabilities to the meter.
  • Each Orion™ Versa Star Pro meter has four channels that can be customised with interchangeable measurement modules
  • Measurement modules are interchangeable
  • Up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customise measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
Range (pH): −2,000 to +20,000
Range (mV): ±2000,0
Range (Temperature): −5,0 to +105,0 °C
3-year replacement warranty.